Please login to your online account to display your discounted pricing

SHOWA™ Cannonball™ PVC-Coated Gauntlets

Offer outstanding chemical resistance, wear, and flexibility

$4.51 - $5.42

Specifications

Certifications CE
Color Green
Material PVC (Polyvinyl Chloride)
Exterior Finish Rough
For Use With (Application) manufacturing, refining, Petrochemicals, machining, construction, utilities, maintenance
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity    

28-705-05

 
showa™
81410
14 in. 35.5cm Large Pair for $5.42
Case of 72 Pair for $352.80

28-705-03

 
showa™
L81108
11 in. 27.9cm Small Pair for $4.83
Case of 72 Pair for $313.92

11-395-4

 
showa™
81210
12 in. 30.5cm Large Pair for $4.51
Case of 72 Pair for $293.76
Description & Specifications

Specifications

Certifications CE
Color Green
Material PVC (Polyvinyl Chloride)
Exterior Finish Rough
For Use With (Application) manufacturing, refining, Petrochemicals, machining, construction, utilities, maintenance
Glove Cuff Style Gauntlet
Lining Jersey

Ideal for refining, construction, mining, utilities, and handling petrochemicals or abrasive materials coated with oil or grease.

  • Feature a nonslip grip and a high-grade PVC plastic coating
  • Cotton jersey liner enhances worker comfort and dexterity
  • Sanitized™
  • Color: Forest green

Recyclable. Made in U.S.A.