Please login to your online account to display your discounted pricing

SHOWA™ Cannonball™ PVC-Coated Gauntlets


Offer outstanding chemical resistance, wear, and flexibility

Manufacturer: showa™  81210

 View more versions of this product

Catalog No. 11-395-4

  • / Pair
  • / Case of 72 Pair
 

Description & Specifications

Specifications

Certifications CE
Length (Metric) 30.5cm
Exterior Finish Rough
Glove Cuff Style Gauntlet
For Use With (Application) manufacturing, refining, Petrochemicals, machining, construction, utilities, maintenance
Length (English) 12 in.
Lining Jersey
Material PVC
Size Large
Color Green