Please login to your online account to display your discounted pricing

SHOWA™ Butyl Gloves


Ideal for use with aldehydes, ketones and esters

Manufacturer: showa™  878XXL11

 View more versions of this product

Catalog No. 11-395-5C

  • / Pair
 

Description & Specifications

Specifications

Size 11
Certifications EN 388
Glove Cuff Style Rolled
For Use With (Application) manufacturing handling Ketones, Alcohols, nerve agents, military, homeland security, Level A suits, hazmat
Liner Material Unlined
Exterior Finish Smooth
Material Butyl
Length (Metric) 35.5cm
Color Black
Thickness 25mil
Length (English) 14 in.