Please login to your online account to display your discounted pricing

SHOWA™ Butyl Gloves


Ideal for use with aldehydes, ketones and esters

Manufacturer: showa™  878XL10

 View more versions of this product

Catalog No. 11-395-5B

  • / Pair
 

Description & Specifications

Specifications

Size 10
Certifications EN 388
For Use With (Application) manufacturing handling Ketones, Alcohols, nerve agents, military, homeland security, Level A suits, hazmat
Glove Cuff Style Rolled
Liner Material Unlined
Exterior Finish Smooth
Length (Metric) 35.5cm
Material Butyl
Color Black
Thickness 25mil
Length (English) 14 in.