missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SHOWA™ Heavy Duty PVC Coated Seamless Knit Jersey Gloves
PVC coating is breathable while also minimizing the absorption of water, oil, and grease. Cotton jersey liner ensures a soft, lightweight fit.
$6.02 - $6.35
Specifications
Coated Area | Full Glove |
---|---|
Construction | Seamless Knit |
Coating Material | PVC |
Color | Yellow |
Exterior Finish | Fully Textured |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
11-388-131C
|
SHOWA™
962L10 |
Large |
Pair for $6.02
Case of 72 Pair for $403.92
|
|
|||||
11-388-131B
|
SHOWA™
962M09 |
Medium |
Pair for $6.35
Case of 72 Pair for $412.56
|
|
|||||
Description
- PVC coating ensures breathability
- Easy donning and doffing with slip on design
- Cotton jersey liner is soft and lightweight
- Outer layer minimizes absorption of water, oil, and grease
- Texturized grip for wet or oily applications
Specifications
Full Glove | |
PVC | |
Fully Textured | |
Seamless Knit Gloves | |
Open | |
962 |
Seamless Knit | |
Yellow | |
Specific | |
Landscaping, Utility Work, and Wood Cutting | |
Cotton Jersey |