Please login to your online account to display your discounted pricing

SHOWA™ Fuzzy Duck™ Gloves


Cool and abrasion resistant

Manufacturer: showa™  962M09

 View more versions of this product

Catalog No. 11-388-131B

  • / Pair
  • / Case of 72 Pair
 

Description & Specifications

Specifications

Cleaning Procedure Launder at 40°C using a neutral Detergent
Exterior Finish Textured
Glove Cuff Style Slip-on
For Use With (Application) small parts handling, general maintenance, shipping and assembly
Material PVC
Liner Material Cotton Jersey
Size 9
Coated Area Full
Color Yellow
Cut Resistance Rating Level 1