missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SHOWA™ CleaN-DEX™ Ultimate 100% Nitrile Gloves

Ideal for hand protection in critical environments

Manufacturer:  SHOWA™ C9905PFXL*

 View more versions of this product

Catalog No. 191301565D




Description

Description

Class 1000 clean room.

  • Clean-processed 100% Nitrile glove
  • Delivers exceedingly good ESD properties and particulate counts, protecting high-end semi-conductors and perishable foodstuffs alike
  • “Clean” white color is ideal for clean room, lab and food processing environments that require exacting standards of industrial hygiene
  • Textured fingertips provide enhanced grip and tactile discrimination
  • Long, beaded cuff design ensures compatibility with clean room garments and apparel

  • Cleanroom operations
  • Food processing
  • Electronics manufacturing
  • Pharmaceuticals
  • Laboratories
  • Intricate parts handling
  • Paint applications

Order Info

Packed 100 gloves per polybag, then double-bagged, then placed in master case polyliner to ensure integrity. Manufactured in an ISO 9002-certified facility. 12 in. long (30.5cm), 6mil thickness.

Compliance

Comply with 21CFR 170-199.

Specifications

Specifications

Textured
Ambidextrous
X-Large
Powder-free
12 in.
Long, Beaded
Nitrile
6mil
White
30.48cm
Documents