missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SHOWA™ C9905PF Single Use Nitrile Cleanroom Gloves

Silicon free, clean-processed highly chemical resistant cleanroom glove with high tear resistance and excellent grip!

$59.63 - $65.90

Specifications

Color White
Length (English) 12 in.
Length (Metric) 30 cm
Material Nitrile
Exterior Finish Textured finger-tips
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-130-1565C
View Documents
SHOWA™
C9905PFLW
Large
Pack of 100 Each for $65.90
Case of 10 Pack for $631.10
 
191301565B
View Documents
SHOWA™
C9905PFM*
Medium
Pack of 100 Each for $59.63
Case of 10 Pack for $571.10
 
19-048656
View Documents
SHOWA™
C9905PFS
Small
Pack of 100 Each for $62.32
Case of 10 Pack for $596.90
 
191301565D
View Documents
SHOWA™
C9905PFXL*
X-Large
Pack of 100 Each for $59.63
Case of 10 Pack for $571.10
 
191301565
View Documents
SHOWA™
C9905PFXS*
X-Small
Pack of 100 Each for $59.63
Case of 10 Pack for $571.10
 
Description

Description

Class 100 cleanroom (ISO 5).

  • Clean-processed 100% Nitrile glove.
  • Delivers exceedingly good ESD properties and particulate counts, protecting high-end semi-conductors and perishable foodstuffs alike.
  • White color is ideal for clean room, lab and food processing environments that require exacting standards of industrial hygiene.
  • Textured finger-tips provide enhanced grip and tactile discrimination.
  • Long, beaded cuff design ensures compatibility with clean room garments and apparel.
  • Category III, EN 374 approved.

  • Cleanroom operations.
  • Food processing.
  • Electronics manufacturing.
  • Pharmaceuticals.
  • Laboratories.
  • Intricate parts handling.
  • Paint applications.

Order Info

Packed 100 gloves per polybag, then double-bagged, then placed in master case polyliner to ensure integrity. Manufactured in an ISO 9002-certified facility. 12 in. long (30.5cm), 6mil thickness.

Compliance

Comply with 21CFR 170-199.

Specifications

Specifications

White
30 cm
Textured finger-tips
Powder-free
12 in.
Nitrile
Long, Beaded
5 mil
SDS