missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SHOWA™ 874 Best™ Butyl II Gloves
Same chemical permeation resistance
Manufacturer: SHOWA™ 87411
Description
The gloves are made with thinner construction for better dexterity.
- Available in both smooth and rough finish
Specifications
14 mil | |
Rolled | |
Butyl II | |
11 | |
14 in. | |
EN 388 |
Smooth | |
manufacturing handling Ketones, Alcohols, nerve agents, military, homeland security, Level A suits, hazmat | |
Unlined | |
Black | |
35.5 cm |