missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SHOWA™ 874 Series Chemical Resistant Butyl Gloves

Unsupported butyl chemical resistant gloves offer protection against highly corrosive acids, ketones, and esters.

Manufacturer:  SHOWA™ 874R10

 View more versions of this product

Catalog No. 19050072D



Description

Description

  • Butyl rubber
  • High permeation resistance to gases and water vapors
  • Unlined, rolled cuff
  • Available in smooth or rough grip
  • Unsupported
  • Thin construction for better dexterity
Specifications

Specifications

Black
Rolled
Unlined
874R-10
14 in.
Cat III, CE 0299
EN 388:0010X
BIK
Yes
14 mil
Rough
Manufacturing, chemical handling, nerve agents, military, homeland security, hazmat
Unlined
Butyl
10
35.5 cm
EN 388
Type B
Bacteria, Fungi WITH Virus Protection
874 Series
0.35 mm
SDS
Documents
Product Certifications