Please login to your online account to display your discounted pricing
SHOWA™ Answer™ and Answer Plus™ Gloves
Combine the look and feel of leather with the durability and flexibility of a nitrile-coated fabric
$4.30 - $4.96
Products
Catalog Number Mfr. No. Price Quantity    
28-708-98
showa™
273508
Pair for $4.30
Case of 144 Pair for $551.52
11-395-6B
showa™
273507
Pair for $4.96
Case of 144 Pair for $637.92
Description & Specifications
  • Both styles feature special nitrile compound laminated onto exterior of cotton-poly base; the result is extra strength
  • Unique patterned finish provides excellent wet and dry grip
  • Sanitized™ interior, an antimicrobial, bacteriostatic treatment that does not wash out with laundering, extends the life of the gloves
  • Perforated back of Answer Plus version keeps hands extra cool
  • Talc-free to avoid silica contamination
  • Launderable
Compliant with 21CFR170-199.