Please login to your online account to display your discounted pricing

BES (White Crystalline Powder), Fisher BioReagents


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLBP501500

 View more versions of this product

Catalog No. BP501-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Melting Point 152°C
Formula Weight 213.25
Quantity 500g
Physical Form Solid
pH 2.5 to 5.0
Color White
Molecular Formula C6H15NO5S
DNase DNase free
Protease Protease free
Loss on Drying 1% max.
Packaging Poly Bottle
Health Hazard 2 WARNING!
CAS 10191-18-1
ChemAlert Storage Symbol Gray
pKa 7.0 to 7.2 (at 25°C)
Synonym N, N-bis (2-Hydroxyethyl) Taurine
Assay Percent Range ≥99 %
Health Hazard 3 Emergency Overview
Causes eye, skin, and respiratory tract irritation.
NFPA
Health:1 Flammability:0 Instability:0
Health Hazard 1 Signal Word:WARNING!
Chemical Name or Material BES
Recommended Storage RT

Specifications

Chemical Name or Material BES
CAS 10191-18-1
Physical Form Solid
Assay ≥99%
Assay Percent Range ≥99 %
Molecular Formula C6H15NO5S
Formula Weight 213.25
Synonym N, N-bis (2-Hydroxyethyl) Taurine

  • Used to make BES buffer with a useful pH range of 6.4-7.8 for biological applications
  • Literature references BES as preferred buffer for higher transfection efficiency of plasmid DNA

Accessories
 

Signal Word:WARNING!

Emergency Overview
Causes eye, skin, and respiratory tract irritation.
NFPA
Health:1 Flammability:0 Instability:0

WARNING!

RT

SDS & Safety Documents