missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BES (White Crystalline Powder), Fisher BioReagents

$855.92 - $855.92

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym N, N-bis (2-Hydroxyethyl) Taurine
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanesulfonic acid
SMILES C(CO)N(CCO)CCS(=O)(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP501-500 SDS View Documents Fisher BioReagents
FLBP501500
500g Poly Bottle Each for $855.92
Case of 6 Each for $5,135.52
 
Description

Description

BES is a biological buffer with a usable pH range of 6.4 to 7.8.
Specifications

Chemical Identifiers

10191-18-1
213.248
N, N-bis (2-Hydroxyethyl) Taurine
CHEBI:39045
C(CO)N(CCO)CCS(=O)(=O)O
C6H15NO5S
AJTVSSFTXWNIRG-UHFFFAOYSA-N
73243
2-[bis(2-hydroxyethyl)amino]ethanesulfonic acid

Specifications

10191-18-1
C6H15NO5S
DNase free
C(CO)N(CCO)CCS(=O)(=O)O
213.248
CHEBI:39045
Solid
1% max.
7.0 to 7.2 (at 25°C)
152°C
White
BES
≥99 %
N, N-bis (2-Hydroxyethyl) Taurine
AJTVSSFTXWNIRG-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethanesulfonic acid
73243
213.25
≥99%
Poly Bottle
Protease free
2.5 to 5.0
500g
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Causes eye, skin, and respiratory tract irritation.
NFPA
Health:1 Flammability:0 Instability:0

Signal Word: WARNING!

Recommended Storage : RT