Please login to your online account to display your discounted pricing

BES, Free Acid, ULTROL™ Grade, Calbiochem™, EMD Millipore

Suitable for use in systems where a calcium phosphate-DNA complex formation is desirable

$61.00 - $123.00

Chemical Identifiers

Assay Percent Range ≥99.0%
Formula Weight 213.3
Buffers BES buffer
CAS 10191-18-1
Chemical Name or Material BES, Free Acid
Molecular Formula C6H15NO5S
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

391334100GM

 
emd millipore corporation
391334100GM
Each for $123.00

39133425GM

 
emd millipore corporation
39133425GM
Each for $61.00
Description & Specifications

Specifications

Formula Weight 213.3
Quantity 100g
Purity Grade Notes by titration (dry basis)
Molecular Formula C6H15NO5S
CAS 10191-18-1
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Chemical Name or Material BES, Free Acid
Buffers BES buffer
Assay Percent Range ≥99.0%
Synonym N,N-bis-(2-Hydroxyethyl)-2-aminoethanesulfonic Acid
Solubility Information Partially soluble in cold water
Purity ULTROL™ Grade

Chen and Okayama described an improved transfection system for the stable transformation of cells with plasmid DNA using BES buffer. The gradual formation of the calcium phosphate-DNA complex and its slow precipitation onto cells appears to be the most critical factor contributing to the high efficiency of stable transformations.

Safety and Handling

15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).