missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ BES-buffered saline (2X)

$97.01 - $144.96

Chemical Identifiers

CAS 66992-27-6
Molecular Formula C6H14NNaO5S
Molecular Weight (g/mol) 235.23
MDL Number MFCD00217173
InChI Key CFQLQLSIZOWFNV-UHFFFAOYSA-M
PubChem CID 23671640
IUPAC Name sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
SMILES C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61955AK View Documents Alfa Aesar™
J61955AK
250mL Each for $97.01
 
AAJ61955AP View Documents Alfa Aesar™
J61955AP
500mL Each for $144.96
 
Specifications

Chemical Identifiers

66992-27-6
235.23
CFQLQLSIZOWFNV-UHFFFAOYSA-M
sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
C6H14NNaO5S
MFCD00217173
23671640
C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]

Specifications

66992-27-6
MFCD00217173
CFQLQLSIZOWFNV-UHFFFAOYSA-M
sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
23671640
250mL
C6H14NNaO5S
2X
C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]
235.23
Liquid
BES-buffered saline