Please login to your online account to display your discounted pricing

Alfa Aesar™ Beryllon II

$52.96 - $52.96

Chemical Identifiers

Chemical Name or Material Beryllon II
CAS 51550-25-5
Molecular Formula C20H10N2Na4O15S4
Formula Weight 738.52
MDL Number MFCD00046409
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2438706

 
alfa aesar™
B2438706
5g Each for $52.96
Description & Specifications

Specifications

Formula Weight 738.52
Chemical Name or Material Beryllon II
TSCA TSCA
Molecular Formula C20H10N2Na4O15S4
MDL Number MFCD00046409
CAS 51550-25-5
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.