missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Beryllium sulfate tetrahydrate, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1610414

 View more versions of this product

Catalog No. AA1610414



Specifications

Specifications

Beryllium sulfate tetrahydrate
(metals basis)
MFCD00149156
14,1175
Freely soluble in water; Insoluble in alcohol
[Be+2].O.O.O.O.[O-]S(=O)(=O)[O-]
177.128
CHEBI:53502
Lumps
1.1713g/mL
7787-56-6
BeH8O8S
UN1566
beryllium sulfate tetrahydrate, ccris 85, beryllium sulphate tetrahydrate, beryllium monosulfate tetrahydrate, unii-2tyk3lf8zn, beryllium sulfate, tetrahydrate 1:1:4, 2tyk3lf8zn, sulfuric acid, beryllium salt 1:1 , tetrahydrate, beryllium sulfate tetrahydrate beryllium and beryllium compounds, dsstox_cid_4603
DIMYTQPLZWDZFE-UHFFFAOYSA-L
beryllium;sulfate;tetrahydrate
62672
177.14 (105.07 Anhydrous)
99.99%
25g

Chemical Identifiers

7787-56-6
177.128
DIMYTQPLZWDZFE-UHFFFAOYSA-L
62672
beryllium;sulfate;tetrahydrate
BeH8O8S
MFCD00149156
beryllium sulfate tetrahydrate, ccris 85, beryllium sulphate tetrahydrate, beryllium monosulfate tetrahydrate, unii-2tyk3lf8zn, beryllium sulfate, tetrahydrate 1:1:4, 2tyk3lf8zn, sulfuric acid, beryllium salt 1:1 , tetrahydrate, beryllium sulfate tetrahydrate beryllium and beryllium compounds, dsstox_cid_4603
CHEBI:53502
[Be+2].O.O.O.O.[O-]S(=O)(=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BERYLLIUM COMPOUNDS, N.O.S.

TSCA : TSCA

SDS Documents