Please login to your online account to display your discounted pricing

Beryllium powder, -325 mesh, 99+% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Beryllium powder
CAS 7440-41-7
MDL Number MFCD00134032
UN Number UN1567
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1010806

  SDS
alfa aesar
1010806
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Beryllium powder
TSCA TSCA
Assay ≥99%
Assay Percent Notes (metals basis)
MDL Number MFCD00134032
CAS 7440-41-7
UN Number UN1567
Mesh Size −325 mesh
Odor Odorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335;
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BERYLLIUM, POWDER
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335;
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.