Learn More
Alfa Aesar™ Beryllium, plasma standard solution, Specpure™, Be 10,000μg/mL
Manufacturer: Alfa Aesar™ 14406AE
Specifications
Beryllium plasma standard solution | |
MFCD00134032 | |
Be4O(C2H3O2)6 in 5% HNO3 | |
Specpure™ |
Be 10000μg/mL | |
UN2922 | |
Liquid | |
100mL |
Chemical Identifiers
MFCD00134032 |
Safety and Handling
GHS H Statement
H350-H372-H314-H302-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.