missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Beryllium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03548006

 View more versions of this product

Catalog No. AA3548006


Description

Description

Beryllium oxide is used in rocket engines. It is also used as a structural ceramic for high-performance microwave devices, vacuum tubes, magnetrons, and gas lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4300°C
2575°C
Powder
(metals basis)
MFCD00003457
14,1172
Insoluble in water.
[Be]=O
25.01
CHEBI:62842
99.95%
3.01 g/mL
Beryllium oxide
1304-56-9
5 g
BeO
UN1566
beryllium oxide, beryllia, bromellete, thermalox, beryllium monoxide, natural bromellite, thermalox 995, ccris 83, beryllium oxide beryllium and beryllium compounds
LTPBRCUWZOMYOC-UHFFFAOYSA-N
oxoberyllium
14775
25.01
Odorless

Chemical Identifiers

1304-56-9
25.01
LTPBRCUWZOMYOC-UHFFFAOYSA-N
14775
oxoberyllium
BeO
MFCD00003457
beryllium oxide, beryllia, bromellete, thermalox, beryllium monoxide, natural bromellite, thermalox 995, ccris 83, beryllium oxide beryllium and beryllium compounds
CHEBI:62842
[Be]=O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P271-P280i-P281-P284-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P501c

H301-H315-H319-H330-H334-H335-H350i-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BERYLLIUM COMPOUNDS, N.O.S.

EINECSNumber : 215-133-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only