missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Beryllium lump, irregular, 3 to 38mm (0.1 to 1.5 in.), 99.5% (metals basis), Alfa Aesar™
Manufacturer: Alfa Aesar 1285309
Specifications
Beryllium | |
7440-41-7 | |
99.5% (Metals basis) | |
MFCD00134032 | |
ATBAMAFKBVZNFJ-UHFFFAOYSA-N | |
beryllium | |
9.012 | |
CHEBI:33783 | |
Gray |
10g | |
Gray irregular lump | |
Be | |
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds | |
[Be] | |
Odorless | |
5460467 | |
Irregular Lump, 3-38mm (0.1-1.5 In.) |
Chemical Identifiers
7440-41-7 | |
9.012 | |
ATBAMAFKBVZNFJ-UHFFFAOYSA-N | |
5460467 | |
beryllium |
Be | |
MFCD00134032 | |
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds | |
CHEBI:33783 | |
[Be] |
Safety and Handling
H301-H330-H315-H319-H317-H335-H350-H372
EINECSNumber : 231-150-7