missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Beryllium ICP Standard, 1000 ppm Be in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PBE1KN100
Specifications
Beryllium ICP Standard | |
Natural LDPE Bottle | |
91.0,6.01,1.03 | |
Colorless | |
1 mL = 1 mg Be |
19049-40-2,7697-37-2,19049-40-2 | |
100 mL | |
95.0,6.26,1.07 | |
1,000ppm Be | |
Liquid |
Chemical Identifiers
19049-40-2 |