missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Beryllium ICP Standard, 1mL = 10mg Be (10,000ppm Be), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPBE10KN50

 View more versions of this product

Catalog No. PBE10KN-50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Beryllium ICP Standard
70.0,20.01,7.9
Liquid
10,000ppm Be
Colorless
19049-40-2,7697-37-2,19049-40-2
73.0,20.83,8.23
Poly natural
1 mL = 10 mg Be
50mL

Chemical Identifiers

19049-40-2
Documents