missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Beryllium foil, 0.5mm (0.02 in.) thick, 99.4% (metals basis)

Manufacturer:  Alfa Aesar™ 14756FI

 View more versions of this product

Catalog No. AA14756FI



Specifications

Specifications

Beryllium foil
50 x 50mm
7440-41-7
(metals basis)
MFCD00134032
Odorless
beryllium
5460467
Foil
≈0.6g/25x25mm
0.5mm (0.02 in.) thick, Vacuum tight
99.4%
Be
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
[Be]
9.012
CHEBI:33783

Chemical Identifiers

7440-41-7
9.012
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
CHEBI:33783
[Be]
Be
MFCD00134032
5460467
beryllium
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

SDS Documents