Please login to your online account to display your discounted pricing

Beryllium 2,4-pentanedionate, White powder, 99%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Beryllium 2, 4-pentanedionate
CAS 10210-64-7
Physical Form Powder
Assay Percent Range 99%
Molecular Formula C10H14BeO4
Formula Weight 207.23
MDL Number MFCD00013485
Synonym Beryllium acetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3857514

 
alfa aesar
3857514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3857522

 
alfa aesar
3857522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 207.23
Quantity 25g
Color White
DOT Information DOT Class: 6.1, Packaging Group: II
Chemical Name or Material Beryllium 2, 4-pentanedionate
EINECS Number 233-513-5
Odor Odorless
Assay Percent Range 99%
Molecular Formula C10H14BeO4
MDL Number MFCD00013485
CAS 10210-64-7
Synonym Beryllium acetylacetonate
Health Hazard 1 H301-H330-H315-H319-H317-H335-H350-H372-H411: Toxic if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.
Safety and Handling

H301-H330-H315-H319-H317-H335-H350-H372-H411: Toxic if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.