Berkshire™ Twillx™ 1622 - ISO Class 6
For ISO Class 6 (Class 1000) cleanroom applications
$184.90 - $184.90
Specifications
Autoclavable Yes
Basis Weight 171
Certifications/Compliance RoHS 2002/05, REACH
Compatibility IPA and other solvents
For Use With (Equipment) For high temperature equipment cleaning and diffusion furnaces
View More Specs
Products 1
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
17-987-88C View Documents Berkshire™
T162209098
Pack of 300 Each for $184.90
Case of 8 Pack for $1,479.20
Description
Description
  • Made of 100% cotton twill with bias-cut edges to eliminate potential raveling
  • Natural fibers provide excellent absorbency, good thermal stability and are highly strong and durable
  • Ideal for cleaning, burnishing and polishing rigid disks, and cleaning grooved, slotted and hard-to-reach places
  • Also good for heavy-duty wiping and cleanup on diffusion furnaces
  • 100% biodegradable
Order Info
Basis weight: 171g/m2
Specifications
Specifications
Yes
RoHS 2002/05, REACH
For high temperature equipment cleaning and diffusion furnaces
Cotton
171
IPA and other solvents
For cleaning, polishing and burnishing of Metallic and nonmetallic magnetic media disk surfaces
Twillx™ 1622 wipes