missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Berkshire™ Durx™ 770 Wipers

For ISO Class 5 (Class 100) applications

Manufacturer:  Berkshire™ DR770121220

 View more versions of this product

Catalog No. 06-665-33A



Description

Description

  • Made from mechanically hydroentangled 55% cellulose/45% polyester with proprietary non-ionic binder
  • Work well with IPA and other solvents
  • Low fiber and particle generation
  • Soft, highly sorbent and durable
  • Critical combination of synthetic polyester strength with the absorbent characteristics of cellulose

Order Info

Basis weight, 65.8g/m2.

Specifications

Specifications

12 in.
Yes
RoHS
Common cleaning and disinfecting solutions
Cleaning of lab equipment, integrated circuit, printed circuit board and electronic components
30.48 cm
Durx™ 770 cleanroom wipes
30.48 cm
65.8
White
12 x 12 in. (30.48 x 30.48 cm)
12 in.
Cellulose, Polyester
SDS
Documents