Berkshire™ BCR™ Cleanroom Masks
Designed to protect cleanrooms and controlled environment from human contamination
Manufacturer:  Berkshire™ BM112*
 View more versions of this product
Catalog No. 01-378
Description
Description
Masks contain no surfactants, fiberglass, glue, binders, or thread.
  • Provide superior filtration efficiency and long wearing comfort for operators
  • Soft, comfortable inside layer provides extremely efficient particle and bacteria filtration and excellent breathability measured in delta P
  • Hems, seams, and attachments are ultrasonically bound for strength and cleanliness
  • Cat. No 01-378 is individually wrapped and includes a clip that allows mask to be worn behind the head
  • Cat. No 19-035-071 comes bulk packaged and is extra wide reducing the amount of exposed skin
Specifications
Specifications
Safety
3-ply construction, Polypropylene outerfacing, non-shedding Polyurethane earloops, adjustable Vinyl covered Steel fully enclosed nose piece, ultrasonically bound hems, seams and attachments
PP (Polypropylene)
18cm
Blue
Knitted Nylon earloops, Polyethylene clip
7.08 in.
Documents