Learn More
Benzyltriphenylphosphonium chloride, 99%
CAS: 1100-88-5 | C25H22ClP | 388.87 g/mol
Supplier: Thermo Scientific Chemicals A1403422
Description
Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.
Solubility
Soluble in water and alcohol. Insoluble in acetone.
Notes
Hygroscopic. Store under inert gas. Store away from strong oxidizing agents, water/ moisture..
Specifications
| Benzyltriphenylphosphonium chloride | |
| ∼330°C (decomposition) | |
| Odorless | |
| MFCD00011913 | |
| UN3464 | |
| Hygroscopic | |
| Soluble in water and alcohol. Insoluble in acetone. | |
| [Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 388.87 | |
| 388.88 |
| 1100-88-5 | |
| >300°C (572°F) | |
| C25H22ClP | |
| 100 g | |
| 3599868 | |
| benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid | |
| USFRYJRPHFMVBZ-UHFFFAOYSA-M | |
| benzyl(triphenyl)phosphanium;chloride | |
| 70671 | |
| 99% |
Chemical Identifiers
| 1100-88-5 | |
| 388.87 | |
| USFRYJRPHFMVBZ-UHFFFAOYSA-M | |
| 70671 | |
| [Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C25H22ClP | |
| MFCD00011913 | |
| benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid | |
| benzyl(triphenyl)phosphanium;chloride |
Safety and Handling
GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H300+H330-H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
EINECSNumber : 214-154-3
RTECSNumber : TA2416500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only