missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltriphenylphosphonium Chloride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 106215000

Catalog No. AC106215000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For C-C Bond Formation (Olefination)
Specifications

Specifications

Benzyltriphenylphosphonium chloride
98.5
>300°C
98.5% min. (Argentometry)
C25H22ClP
MFCD00011913
benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid
USFRYJRPHFMVBZ-UHFFFAOYSA-M
benzyl(triphenyl)phosphanium;chloride
70671
99%
1100-88-5
100.0
Authentic
Glass bottle
C6H5CH2P(C6H5)3Cl
500g
Solubility in water: soluble. Other solubilities: soluble in chloroform,slightly soluble in lower aliphatic alcohols
[Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
388.87
388.86
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with wat

GHS Signal Word: Danger

EINECSNumber : 214-154-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only