Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyltriphenylphosphonium chloride, 99%

Chemical Identifiers

Chemical Name or Material Benzyltriphenylphosphonium chloride
CAS 1100-88-5
Molecular Formula C6H5CH2P(C6H5)3Cl
Formula Weight 388.88
MDL Number MFCD00011913
UN Number UN3464
Beilstein 3599868
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1403422

 
alfa aesar™
A1403422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1403436

 
alfa aesar™
A1403436
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 388.88
RTECS Number TA2416500
TSCA TSCA
Chemical Name or Material Benzyltriphenylphosphonium chloride
MDL Number MFCD00011913
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
Molecular Formula C6H5CH2P(C6H5)3Cl
Quantity 100g
Odor Odorless
Melting Point ≈330°C (dec.)
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
UN Number UN3464
CAS 1100-88-5
Beilstein 3599868
Assay 99%
Flash Point >300°C (572°F)
Health Hazard 2 GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.

Solubility
Soluble in water and alcohol. Insoluble in acetone.

Notes
Hygroscopic. Store under inert gas. Store away from strong oxidizing agents, water/ moisture..