missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylsilane, TCI America™

$48.89 - $151.25

Chemical Identifiers

CAS 770-09-2
Molecular Formula C10H16Si
Molecular Weight (g/mol) 164.323
MDL Number MFCD00039796
InChI Key MRIWRLGWLMRJIW-UHFFFAOYSA-N
Synonym trimethylbenzylsilane, silane, trimethyl phenylmethyl, silane, benzyltrimethyl, alpha-trimethylsilyltoluene, benzyl trimethyl silane, unii-hfx8b6q8wc, hfx8b6q8wc, benzene, trimethylsilyl methyl, benzene, trimethylsilyl methyl, amtsi018
PubChem CID 69862
IUPAC Name benzyl(trimethyl)silane
SMILES C[Si](C)(C)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B18565ML View Documents TCI America
B18565ML
5mL Each for $48.89
 
B185625ML View Documents TCI America
B185625ML
25mL Each for $151.25
 
Specifications

Chemical Identifiers

770-09-2
164.323
MRIWRLGWLMRJIW-UHFFFAOYSA-N
69862
C[Si](C)(C)CC1=CC=CC=C1
C10H16Si
MFCD00039796
trimethylbenzylsilane, silane, trimethyl phenylmethyl, silane, benzyltrimethyl, alpha-trimethylsilyltoluene, benzyl trimethyl silane, unii-hfx8b6q8wc, hfx8b6q8wc, benzene, trimethylsilyl methyl, benzene, trimethylsilyl methyl, amtsi018
benzyl(trimethyl)silane

Specifications

191°C
5mL
C10H16Si
1993
MRIWRLGWLMRJIW-UHFFFAOYSA-N
benzyl(trimethyl)silane
69862
Clear Liquid at 20°C
Benzyltrimethylsilane
Colorless
770-09-2
MFCD00039796
trimethylbenzylsilane, silane, trimethyl phenylmethyl, silane, benzyltrimethyl, alpha-trimethylsilyltoluene, benzyl trimethyl silane, unii-hfx8b6q8wc, hfx8b6q8wc, benzene, trimethylsilyl methyl, benzene, trimethylsilyl methyl, amtsi018
C[Si](C)(C)CC1=CC=CC=C1
164.323
164.32
≥96.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes