Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyltrimethylsilane, 98%

Chemical Identifiers

Chemical Name or Material Benzyltrimethylsilane
CAS 770-09-2
Molecular Formula C6H5CH2Si(CH3)3
Formula Weight 164.32
MDL Number MFCD00039796
UN Number UN1993
Beilstein 1928113
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1319106

  SDS
alfa aesar™
A1319106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1319122

  SDS
alfa aesar™
A1319122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.32
Assay 98%
MDL Number MFCD00039796
Chemical Name or Material Benzyltrimethylsilane
Molecular Formula C6H5CH2Si(CH3)3
TSCA TSCA
UN Number UN1993
Quantity 5g
Flash Point 57°C (134°F)
Health Hazard 1 Warning
Boiling Point 190° to 191°C
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
CAS 770-09-2
Beilstein 1928113

Applications
Benzyltrimethylsilane is a silicon reagent for generating benzyl anions.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.