missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium Tribromide [Brominating Reagent], TCI America™

Manufacturer:  TCI America T13825G

 View more versions of this product

Catalog No. T13825G



Specifications

Specifications

Benzyltrimethylammonium Tribromide [Brominating Reagent]
102°C
111865-47-5
MFCD00134423
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
benzyl(trimethyl)azanium;molecular bromine;hydrobromide
86222689
Crystal-Powder at 20°C
Undesignated
5g
C10H17Br3N+
benzyltrimethylammonium tribromide
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
390.965
389.96
≥97.0% (T)

Chemical Identifiers

111865-47-5
390.965
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
86222689
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
C10H17Br3N+
MFCD00134423
benzyltrimethylammonium tribromide
benzyl(trimethyl)azanium;molecular bromine;hydrobromide
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2694

TSCA : Yes

Documents