missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyltrimethylammonium tribromide, 98%

$32.46 - $341.07

Chemical Identifiers

CAS 111865-47-5
Molecular Formula C6H5CH2N(CH3)3Br3
Formula Weight 389.97
MDL Number MFCD00134423
Chemical Name or Material Benzyltrimethylammonium tribromide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2447003 SDS Alfa Aesar™
B2447003
1g Each for $32.46
AAB2447006 SDS Alfa Aesar™
B2447006
5g Each for $99.62
AAB2447014 SDS Alfa Aesar™
B2447014
25g Each for $341.07
Specifications Description & Specifications

Chemical Identifiers

111865-47-5
389.97
Benzyltrimethylammonium tribromide
C6H5CH2N(CH3)3Br3
MFCD00134423

Specifications

111865-47-5
389.97
99° to 101°C
Benzyltrimethylammonium tribromide
C6H5CH2N(CH3)3Br3
MFCD00134423
1g
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning