missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium Methoxide, 40 wt. %solution in Methanol, ACROS Organics™

Manufacturer:  Acros Organics 156500010

 View more versions of this product

Catalog No. AC156500010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Phase transfer catalyst
Specifications

Specifications

Benzyltrimethylammonium methoxide
122-08-7,67-56-1
41.0,61.0
C6H5CH2N(CH3)3OCH3
benzyltrimethylammonium methoxide, n,n,n-trimethylbenzenemethanaminium methoxide, benzyltrimethylammonium methanolate, benzenemethanaminium, n,n,n-trimethyl-, methoxide, benzyltrimethylazanium; methoxide, trimethyl benzyl ammonium methoxide, ammonium, benzyltrimethyl-, methoxide, benzenemethanaminium, n,n,n-trimethyl-, salt with methanol 1:1, c10h16n.ch3o, ammonium, methoxide
C[N+](C)(C)CC1=CC=CC=C1.C[O-]
181.279
181.28
≥39%
Colorless to Yellow
40 wt. %solution in Methanol
39.0,59.0
C11H19NO
MFCD00011783
SMTUJUHULKBTBS-UHFFFAOYSA-N
benzyl(trimethyl)azanium;methanolate
8504
Solution
Glass bottle
1L

Chemical Identifiers

122-08-7
181.279
SMTUJUHULKBTBS-UHFFFAOYSA-N
8504
C[N+](C)(C)CC1=CC=CC=C1.C[O-]
C11H19NO
MFCD00011783
benzyltrimethylammonium methoxide, n,n,n-trimethylbenzenemethanaminium methoxide, benzyltrimethylammonium methanolate, benzenemethanaminium, n,n,n-trimethyl-, methoxide, benzyltrimethylazanium; methoxide, trimethyl benzyl ammonium methoxide, ammonium, benzyltrimethyl-, methoxide, benzenemethanaminium, n,n,n-trimethyl-, salt with methanol 1:1, c10h16n.ch3o, ammonium, methoxide
benzyl(trimethyl)azanium;methanolate
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes damage to organs.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-521-6

RTECSNumber : BO8587000

TSCA : TSCA

SDS Documents