missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium Methoxide, 40 wt. %solution in Methanol, ACROS Organics™

Chemical Identifiers

CAS 122-08-7
Molecular Formula C11H19NO
Molecular Weight (g/mol) 181.279
MDL Number MFCD00011783
InChI Key SMTUJUHULKBTBS-UHFFFAOYSA-N
Synonym ammonium, benzyltrimethyl-, methoxide, ammonium, methoxide, benzenemethanaminium, n,n,n-trimethyl-, methoxide, benzenemethanaminium, n,n,n-trimethyl-, salt with methanol 1:1, benzyltrimethylammonium methanolate, benzyltrimethylammonium methoxide, benzyltrimethylazanium; methoxide, c10h16n.ch3o, n,n,n-trimethylbenzenemethanaminium methoxide, trimethyl benzyl ammonium methoxide
PubChem CID 8504
IUPAC Name benzyl(trimethyl)azanium;methanolate
SMILES C[N+](C)(C)CC1=CC=CC=C1.C[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC156500010
SDS View Documents
Thermo Fisher Scientific
156500010
1L Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Phase transfer catalyst
Specifications

Chemical Identifiers

122-08-7
181.279
SMTUJUHULKBTBS-UHFFFAOYSA-N
8504
C[N+](C)(C)CC1=CC=CC=C1.C[O-]
C11H19NO
MFCD00011783
ammonium, benzyltrimethyl-, methoxide, ammonium, methoxide, benzenemethanaminium, n,n,n-trimethyl-, methoxide, benzenemethanaminium, n,n,n-trimethyl-, salt with methanol 1:1, benzyltrimethylammonium methanolate, benzyltrimethylammonium methoxide, benzyltrimethylazanium; methoxide, c10h16n.ch3o, n,n,n-trimethylbenzenemethanaminium methoxide, trimethyl benzyl ammonium methoxide
benzyl(trimethyl)azanium;methanolate

Specifications

122-08-7,67-56-1
41.0,61.0
C6H5CH2N(CH3)3OCH3
ammonium, benzyltrimethyl-, methoxide, ammonium, methoxide, benzenemethanaminium, n,n,n-trimethyl-, methoxide, benzenemethanaminium, n,n,n-trimethyl-, salt with methanol 1:1, benzyltrimethylammonium methanolate, benzyltrimethylammonium methoxide, benzyltrimethylazanium; methoxide, c10h16n.ch3o, n,n,n-trimethylbenzenemethanaminium methoxide, trimethyl benzyl ammonium methoxide
C[N+](C)(C)CC1=CC=CC=C1.C[O-]
181.279
181.28
≥39%
Colorless to Yellow
Benzyltrimethylammonium methoxide, 40 wt. %solution in Methanol
39.0,59.0
C11H19NO
MFCD00011783
SMTUJUHULKBTBS-UHFFFAOYSA-N
benzyl(trimethyl)azanium;methanolate
8504
Solution
Glass bottle
1L
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes damage to organs.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-521-6

RTECSNumber : BO8587000

TSCA : TSCA

SDS