Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyltrimethylammonium iodide, 98%

$69.38 - $183.49

Chemical Identifiers

Chemical Name or Material Benzyltrimethylammonium iodide
CAS 4525-46-6
Molecular Formula C6H5CH2N(CH3)3I
Formula Weight 277.15g/mol
MDL Number MFCD00051859
Beilstein 3917007
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2352014

 
alfa aesar™
B23520-14
25g Each for $69.38

AAB2352022

 
alfa aesar™
B23520-22
100g Each for $183.49
Description & Specifications

Specifications

Melting Point 177° to 182°C
Quantity 25g
Chemical Name or Material Benzyltrimethylammonium iodide
CAS 4525-46-6
Assay 98%
Molecular Formula C6H5CH2N(CH3)3I
Formula Weight 277.15g/mol
MDL Number MFCD00051859
Beilstein 3917007
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number BO8585000
TSCA TSCA