Learn More
Benzyltrimethylammonium hydroxide, 40% w/w in methanol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A14927AE
Description
Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.
Solubility
Miscible with water, hydrocarbons, aromatic hydrocarbons and halogenated solvents.
Notes
Air Sensitive. Incompatible with strong oxidizing agents.
Specifications
Benzyltrimethylammonium hydroxide | |
0.92 | |
15°C (59°F) | |
C10H17NO | |
C6H5CH2N(CH3)3OH | |
100 mL | |
3917256 | |
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide | |
NDKBVBUGCNGSJJ-UHFFFAOYSA-M | |
benzyl(trimethyl)azanium;hydroxide | |
66854 | |
40% w/w in methanol |
100-85-6 | |
65°C | |
Ammonia-like | |
1.427 | |
MFCD00008281 | |
UN2924 | |
Air Sensitive | |
Miscible with water,hydrocarbons,aromatic hydrocarbons and halogenated solvents. | |
C[N+](C)(C)CC1=CC=CC=C1.[OH-] | |
167.252 | |
167.25 |
Chemical Identifiers
100-85-6 | |
167.252 | |
NDKBVBUGCNGSJJ-UHFFFAOYSA-M | |
66854 | |
C[N+](C)(C)CC1=CC=CC=C1.[OH-] |
C10H17NO | |
MFCD00008281 | |
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide | |
benzyl(trimethyl)azanium;hydroxide |
Safety and Handling
GHS H Statement
H225-H311-H331-H370-H314
Highly flammable liquid and vapour.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301+H311+H331-H314-H335-H370-H372
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 202-895-5
RTECSNumber : BO8575000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only