Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyltrimethylammonium hydroxide, 20% w/w aq. soln.

Chemical Identifiers

Chemical Name or Material Benzyltrimethylammonium hydroxide
CAS 100-85-6
Linear Formula C6H5CH2N(CH3)3OH
Formula Weight 167.25
MDL Number MFCD00008281
UN Number UN3267
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41717AD

  SDS
alfa aesar™
41717AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41717AK

  SDS
alfa aesar™
41717AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41717K2

  SDS
alfa aesar™
41717K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.25
Sensitivity Air Sensitive
Color Colorless to pale yellow
Quantity 50mL
Physical Form Liquid
Odor Amine-like
CAS 100-85-6
Linear Formula C6H5CH2N(CH3)3OH
RTECS Number BO8575000
Health Hazard 1 Danger
TSCA TSCA
UN Number UN3267
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00008281
Chemical Name or Material Benzyltrimethylammonium hydroxide
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
Concentration 20% w/w aq. soln.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.