missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium dichloroiodate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 268600250

 View more versions of this product

Catalog No. AC268600250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Benzyltrimethylammonium dichloroiodate
Glass bottle
125°C to 128°C
99%
98.5
99%
C6H5CH2N+(CH3)3Cl2I-
15,25; 08,434; 13,266; 15,278; 16,289; 17,298; 05,571
Solubility in water: soluble in water. Other solubilities: soluble in methylene chloride
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl
2724680
Crystalline Powder or Flakes
Authentic
Yellow to Brown
25g
114971-52-7
100.0
C10H16Cl2IN
MFCD00075259
benzyl trimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, benzyltrimethylammoniumdichloroiodate, benzyltrimethylazanium; dichloroiodanuide, btma-icl2, pubchem21920
PPDJNZTUDFPAHX-UHFFFAOYSA-N
348.04
348.04
99%

Chemical Identifiers

114971-52-7
348.04
PPDJNZTUDFPAHX-UHFFFAOYSA-N
2724680
C10H16Cl2IN
MFCD00075259
benzyl trimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, benzyltrimethylammoniumdichloroiodate, benzyltrimethylazanium; dichloroiodanuide, btma-icl2, pubchem21920
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents