missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium chloride, 50 wt.% solution in water, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 209380010

 View more versions of this product

Catalog No. AC209380010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Benzyltrimethylammonium chloride
50 wt. % Solution in Water
49.0,49.0
C10H16ClN
MFCD00011782
01,53
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
[Cl-].C[N+](C)(C)CC1=CC=CC=C1
185.70
185.7
Glass bottle
1.044
Liquid
Colorless to Yellow
56-93-9,7732-18-5
51.0,51.0
C6H5CH2N(CH3)3Cl
12, 1020
1.0440g/mL
KXHPPCXNWTUNSB-UHFFFAOYSA-M
benzyl(trimethyl)azanium;chloride
5963
≥49%
1.445
1 kg

Chemical Identifiers

56-93-9
185.70
KXHPPCXNWTUNSB-UHFFFAOYSA-M
5963
[Cl-].C[N+](C)(C)CC1=CC=CC=C1
C10H16ClN
MFCD00011782
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
benzyl(trimethyl)azanium;chloride
Safety and Handling

Safety and Handling

GHS P Statement Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF exposed: Call a POISON CENTRE or doctor/physician. Avoid release to the environment.

Danger

EINECSNumber : 200-300-3

RTECSNumber : BO8400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only