Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyltriethylammonium tetrafluoroborate, 98+%

$64.34 - $233.55

Chemical Identifiers

Chemical Name or Material Benzyltriethylammonium tetrafluoroborate
CAS 77794-93-5
Molecular Formula (CH3CH2)3N(CH2C6H5)BF4
Formula Weight 279.13
MDL Number MFCD00011823
Beilstein 5198002
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2484709

  SDS
alfa aesar™
B2484709
10g Each for $64.34

AAB2484718

  SDS
alfa aesar™
B2484718
50g Each for $233.55
Description & Specifications

Specifications

Formula Weight 279.13
Chemical Name or Material Benzyltriethylammonium tetrafluoroborate
Assay ≥98%
Molecular Formula (CH3CH2)3N(CH2C6H5)BF4
MDL Number MFCD00011823
CAS 77794-93-5
Beilstein 5198002
Melting Point 113° to 115°C
Health Hazard 1 Warning
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.