Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyltriethylammonium dichloroiodate, 97%

Chemical Identifiers

Chemical Name or Material Benzyltriethylammonium dichloroiodate
Molecular Formula C13H22Cl2IN
Linear Formula (CH3CH2)3N(CH2C6H5)ICl2
Formula Weight 390.13
MDL Number MFCD11973847
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5187203

  SDS
alfa aesar™
H5187203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5187206

  SDS
alfa aesar™
H5187206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5187214

  SDS
alfa aesar™
H5187214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 390.13
Chemical Name or Material Benzyltriethylammonium dichloroiodate
Assay 97%
Molecular Formula C13H22Cl2IN
Linear Formula (CH3CH2)3N(CH2C6H5)ICl2
MDL Number MFCD11973847
Melting Point 79° to 81°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.