Learn More
Benzyltriethylammonium chloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC164020025
Description
Phase transfer catalystSpecifications
Benzyltriethylammonium chloride | |
Authentic | |
1% max. (K.F.) | |
185.0°C | |
98% | |
97.5 | |
97.5 to 102.5% (Argentometry) | |
C6H5CH2N(C2H5)3Cl | |
benzyl triethyl ammonium chloride, benzyl triethylammonium chloride, benzyltriethylammonium chloride, benzyltriethylammoniumchloride, benzyltriethylazanium chloride, bteac, n-benzyl-n,n-diethylethanaminium chloride, teba, tebac, triethylbenzylammonium chloride | |
HTZCNXWZYVXIMZ-UHFFFAOYSA-M | |
benzyl(triethyl)azanium;chloride | |
66133 | |
Crystalline Powder |
>275°C | |
Plastic bottle | |
White to Beige | |
2.5kg | |
56-37-1 | |
100.0 | |
C13H22ClN | |
MFCD00011824 | |
Solubility in water: 700g/L water (20°C). Other solubilities: soluble in methanol,ethanol,isopropanol,dmf,soluble in acetone,ch2cl2 | |
CC[N+](CC)(CC)CC1=CC=CC=C1.[Cl-] | |
227.78 | |
227.78 | |
98% |
Chemical Identifiers
56-37-1 | |
227.78 | |
HTZCNXWZYVXIMZ-UHFFFAOYSA-M | |
66133 | |
CC[N+](CC)(CC)CC1=CC=CC=C1.[Cl-] |
C13H22ClN | |
MFCD00011824 | |
benzyl triethyl ammonium chloride, benzyl triethylammonium chloride, benzyltriethylammonium chloride, benzyltriethylammoniumchloride, benzyltriethylazanium chloride, bteac, n-benzyl-n,n-diethylethanaminium chloride, teba, tebac, triethylbenzylammonium chloride | |
benzyl(triethyl)azanium;chloride |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-270-1
RTECSNumber : BO8275000
TSCA : TSCA