Please login to your online account to display your discounted pricing

Benzyltriethylammonium borohydride, 96%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  302470100

 View more versions of this product

Catalog No. AC302470100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Crystalline Powder
Formula Weight 207.17
Quantity 10g
Color White
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material Benzyltriethylammonium borohydride
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 96%
Molecular Formula C13H26BN
MDL Number MFCD00191785
CAS 85874-45-9
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.

Specifications

Chemical Name or Material Benzyltriethylammonium borohydride
CAS 85874-45-9
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C13H26BN
Formula Weight 207.17
MDL Number MFCD00191785
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.

SDS & Safety Documents