missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltriethylammonium borohydride, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 302470010

 View more versions of this product

Catalog No. AC302470010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Benzyltriethylammonium borohydride
95.0
Authentic
≥95%
C13H26BN
MFCD00191785
1g
85874-45-9
100.0
Crystalline Powder
96%
207.17
White

Chemical Identifiers

85874-45-9
100.0
C13H26BN
MFCD00191785
≥95%
95.0
96%
207.17
Benzyltriethylammonium borohydride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger