missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltriethylammonium borohydride, 96%, ACROS Organics™

Chemical Identifiers

CAS 85874-45-9
Molecular Formula C13H26BN
Molecular Weight (g/mol) 207.17
MDL Number MFCD00191785
InChI Key HQGWCEJYXHFBSA-UHFFFAOYSA-N
Synonym benzyltriethylammonium borohydride, benzyltriethylazanium ? 1-boranuidylidene, n,n,n-triethyl-benzenemethanaminium tetrahydroborate
PubChem CID 10899792
IUPAC Name benzyl(triethyl)azanium;boron(1-)
SMILES [B-].CC[N+](CC)(CC)CC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC302470100
SDS View Documents
Thermo Fisher Scientific
302470100
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Reduction reagent
Specifications

Chemical Identifiers

85874-45-9
207.17
HQGWCEJYXHFBSA-UHFFFAOYSA-N
10899792
[B-].CC[N+](CC)(CC)CC1=CC=CC=C1
C13H26BN
MFCD00191785
benzyltriethylammonium borohydride, benzyltriethylazanium ? 1-boranuidylidene, n,n,n-triethyl-benzenemethanaminium tetrahydroborate
benzyl(triethyl)azanium;boron(1-)

Specifications

Authentic
White
10g
95.0
96%
MFCD00191785
Solubility in water: reacts with water. Other solubilities: soluble in methanol,ethanol,dichloromethane
[B-].CC[N+](CC)(CC)CC1=CC=CC=C1
207.17
207.17
96%
Glass bottle
146°C
85874-45-9
100.0
C13H26BN
benzyltriethylammonium borohydride, benzyltriethylazanium ? 1-boranuidylidene, n,n,n-triethyl-benzenemethanaminium tetrahydroborate
HQGWCEJYXHFBSA-UHFFFAOYSA-N
benzyl(triethyl)azanium;boron(1-)
10899792
Crystalline Powder
Benzyltriethylammonium borohydride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS