missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltributylammonium bromide, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 222892500

 View more versions of this product

Catalog No. AC222892500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
168° to 174°C (dec.)
250g
100.0
Benzyltributylammonium bromide
C19H34BrN
99%
98.5
Authentic
Powder
356.38
White to off-white
Glass bottle
99%
C6H5CH2N[(CH2)3CH3]3Br
25316-59-0
MFCD00011848

Chemical Identifiers

25316-59-0
100.0
C19H34BrN
356.38
Benzyltributylammonium bromide, 99%
98.5
99%
C6H5CH2N[(CH2)3CH3]3Br
MFCD00011848
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 246-819-9