missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltributylammonium bromide, 99%, ACROS Organics™

Chemical Identifiers

CAS 25316-59-0
Molecular Formula C19H34BrN
Molecular Weight (g/mol) 356.38
MDL Number MFCD00011848
InChI Key UDYGXWPMSJPFDG-UHFFFAOYSA-M
Synonym benzenemethanaminium, n,n,n-tributyl-, bromide, benzyl tributyl ammonium bromide, benzyl-tributyl-azanium, benzyltri-n-butylammonium bromide, benzyltributylammonium bromide, benzyltributylammoniumbromide, benzyltributylazanium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, pubchem7801, tributylbenzylammonium bromide
PubChem CID 2724282
IUPAC Name benzyl(tributyl)azanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC222890500
SDS View Documents
Thermo Fisher Scientific
222890500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Phase transfer catalyst
Specifications

Chemical Identifiers

25316-59-0
356.38
UDYGXWPMSJPFDG-UHFFFAOYSA-M
2724282
CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]
C19H34BrN
MFCD00011848
benzenemethanaminium, n,n,n-tributyl-, bromide, benzyl tributyl ammonium bromide, benzyl-tributyl-azanium, benzyltri-n-butylammonium bromide, benzyltributylammonium bromide, benzyltributylammoniumbromide, benzyltributylazanium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, pubchem7801, tributylbenzylammonium bromide
benzyl(tributyl)azanium;bromide

Specifications

Authentic
White
50g
98.5
98.5% min. (Argentometry)
C6H5CH2N[(CH2)3CH3]3Br
benzenemethanaminium, n,n,n-tributyl-, bromide, benzyl tributyl ammonium bromide, benzyl-tributyl-azanium, benzyltri-n-butylammonium bromide, benzyltributylammonium bromide, benzyltributylammoniumbromide, benzyltributylazanium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, pubchem7801, tributylbenzylammonium bromide
CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]
356.38
356.38
99%
Glass bottle
168°C to 174°C
25316-59-0
100.0
C19H34BrN
MFCD00011848
UDYGXWPMSJPFDG-UHFFFAOYSA-M
benzyl(tributyl)azanium;bromide
2724282
Powder
Benzyltributylammonium bromide, 99%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 246-819-9

SDS
Product Certifications